Orders LinkContactUsLinkComponent
Chromatin Extraction Kit, Abcam
Chromatin Extraction Kit, Abcam
Chromatin Extraction Kit
Catalog # ABCAAB117152-100
Marketsource
Supplier:   Abcam
Chromatin Extraction Kit, Abcam
Catalog # ABCAAB117152-100
Supplier:  Abcam
Marketsource
CAS Number:  
Chromatin Extraction Kit, Abcam
Chromatin Extraction Kit
CAS Number:  
Price
8 140,00 Kč
Log in for availability
1 * 100 Tests
Product Details & Documents
60-minute chromatin extraction kit is a complete set of optimized buffers and reagents for isolating chromatin or DNA-protein complex from mammalian cells and tissues in a simple and rapid format. - Cited in over 50 publications
Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications
Specifications
  • Popis:
    Chromatin Extraction Kit
  • Cat. No.:
    ABCAAB117152-100
  • Skladovací teplota:
    Store at Multi.
Specifications