SP Bel-Art Switch-Grid Test Tube Rack, Complete Set, Interchangeable Grids Included
Catalog # BELAF18745-1000
Supplier: Bel-Art Products, a Part of SP
New Product
Specifications
- Pro zkumavky:10 - 30 mm
- Poč. otvorů:12, 24, 40, 60, 84
- Materiál:Polyoxymethylen
- Š×H×V:318×111×670 mm
Specifications
About this item
The innovative Switch-Grid rack utilises interchangeable grids for ultimate flexibility. This split-frame rack holds up to two different tube sizes at the same time and allows users to quickly switch grids to hold other sizes.
10 to 30 mm diameter test tubes fit in 5 color-coded grid inserts
- Can be easily cleaned with common lab detergents, ethanol, or isopropyl alcohol
- Three-tier design securely holds tubes in place while non-slip rubber feet prevent sliding on the lab bench
- Rack available without grids for use with spare grids
This rack is a smart solution for test tube organization and streamlining the lab's workflow. Constructed from chemical-resistant polyoxymethylene plastic, both the frame and grids are autoclavable at 121 °C.