Proteometer-L Full Kit Reagent (1000)
Katalogové číslo NOVIKIT-PL-007-01B
Dodavatel: NOVILYTIC
New Product
Číslo CAS:
Specifikace
- Popis:Proteometer-L full kit reagent (1000)
Specifikace
O tomto produktu
Fluorescent reagent for the Proteometer-L full kit. This is sold separately due to shipping conditions. Store at −20 °C.
- One assay for two critical attributes - titer + aggregate measurement
- No protein A purification required: Reduces costs
- No sample pretreatment: Reduces hours of sample prep
- No method development: Reduces time until method usage
- Direct analysis from CFB: HCP removal not needed
- Plug-and-play for any LC with fluorescence detector: Easy adoption, no software needed
- Low sample volume: Reduces impact on processing
Crimped, sealed, ready for use with the Proteometer-L full kit (NOVIKIT-PL-007-01A) which is purchased separately. This kit is reagent only and pairs with our Proteometer-L full kit.
Reagent to be stored at −20 °C.
Ordering information: Please add Proteometer-L full kit seperately (NOVIKIT-PL-007-01A).
Delivery:
Includes 14× Proteometer-L fluorescent reagent vials - allows for analysis up to 1000 samples.
Packaging: Packaged separately from the Proteometer-UFT full kit due to storage conditions.