Proteometer-CV Full Kit Reagent (300)
Katalogové číslo NOVIKIT-PCV-00301B
Dodavatel: NOVILYTIC
New Product
Číslo CAS:
Specifikace
- Popis:Proteometer-CV full kit reagent (300)
Specifikace
O tomto produktu
Fluorescent reagent portion of the Proteometer-CV full kit. This is sold individually due to shipping and storage conditions.
- No sample preparation required - analysis directly from clarified fermentation broth without Protein A purification
- Speed and efficiency - CV analysis in under 25 minutes
- Simplified workflow - all the components you need for seamless operation
- Exclusive technology - the only solution that performs charge variant analysis directly in clarified fermentation broth
- Reagent to be stored at −20 °C
Crimped, sealed, ready for use with the Proteometer-CV starter kit (NOVIKIT-PCV-00301B), which is purchased separately. This kit is reagent-only and pairs with our Proteometer-CV full kit.
Ordering information: Glassware not included. Please add starter Proteometer-CV kit seperately (NOVIKIT-PCV-00301B).
Delivery: Includes 5× Proteometer-CV fluorescent reagent vials. Allows for analysis up to 300 samples.
Packaging: Packaged separately from the Proteometer-CV Full Kit due to storage conditions.