Proteometer-L Reactor
Catalog # NOVIRCT-PL-001
Supplier: NOVILYTIC
New Product
Specifications
- Popis:Proteometer-L reactor
Specifications
About this item
Replacement reactor for Proteometer-L kit.
- Plugs into any LC with a fluorescence detector
- Store at room temperature
Intended for use with the Proteometer-L kit. Reorder individually as needed.
Packaging: Supplied in a custom box with instructions.