- Popis:Suma Nova Pur-Eco L6 mechanical ware washing detergent for hard water, 2×5 L
- Typ balení:Jerry can
- Bal.:1
- Hodnota pH:≥11
Suma Nova Pur-Eco L6 is a concentrated liquid detergent for mechanical ware washing. It provides excellent soil removal in hard water conditions.
Quickly removes grease and dried-on food
Very effective due to highly concentrated formulation
Reduced impact on aquatic ecosystems
Limited hazardous substances
Performance tested.
Suma Nova Pur-Eco L6 is a heavy duty alkaline liquid detergent suitable for use in single and multi-tank ware washing machines. It contains a blend of alkali and scale control agents that remove fats and dried-on foods. The high level of powerful scale control agents also prevents scaling in hard water conditions.
Suma Nova Pur-Eco L6 is normally applied using Diversey automatic dosing equipment. It can also be used in conjunction with integral dosing pumps where these are fitted.
EU Ecolabel (EU-flower) certified.