ContactUsLinkComponent
Suma® Nova Pur-Eco L6 Mechanical Ware Washing Detergent
Suma® Nova Pur-Eco L6 Mechanical Ware Washing Detergent
  DIVY7515733
 :  Diversey
New Product
Suma® Nova Pur-Eco L6 Mechanical Ware Washing Detergent
  DIVY7515733
 :  Diversey
New Product
  • Popis:
    Suma Nova Pur-Eco L6 mechanical ware washing detergent for hard water, 2×5 L
  • Typ balení:
    Jerry can
  • Bal.:
    1
  • Hodnota pH:
    ≥11
Suma Nova Pur-Eco L6 is a concentrated liquid detergent for mechanical ware washing. It provides excellent soil removal in hard water conditions.
  • Quickly removes grease and dried-on food
    Very effective due to highly concentrated formulation
    Reduced impact on aquatic ecosystems
    Limited hazardous substances
    Performance tested.
Suma Nova Pur-Eco L6 is a heavy duty alkaline liquid detergent suitable for use in single and multi-tank ware washing machines. It contains a blend of alkali and scale control agents that remove fats and dried-on foods. The high level of powerful scale control agents also prevents scaling in hard water conditions.
Suma Nova Pur-Eco L6 is normally applied using Diversey automatic dosing equipment. It can also be used in conjunction with integral dosing pumps where these are fitted.
 :
EU Ecolabel (EU-flower) certified.