0
0,00 Kč
Order Entry
Přihlásit se
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Zadání objednávky
Stav objednávky
Váš účet
Košík
Czech Republic
Zadání objednávky
Stav objednávky
Váš účet
Košík
ContactUsLinkComponent
Domů
Pasti vakuové
Accessories for Cold Trap for Evaporation Tasks, CT50 Single OLÉ
Accessories for Cold Trap for Evaporation Tasks, CT50 Single OLÉ
HUBE504545
:
HUBER
New Product
Accessories for Cold Trap for Evaporation Tasks, CT50 Single OLÉ
HUBE504545
:
HUBER
New Product
Popis:
Glass adaptor, 50 mm flange to GL14 for connection to rotary evaporators
Environmentally Preferable: