Tork T4 Advanced Coreless Conventional Toilet Paper Roll, White
Catalog # SCAA472132
Supplier: Essity
New Product
Specifications
- Balené:24 rolls, 400 sheets each
- Popis:Advanced conventional roll toilet paper, refill, white compatible with T4 dispensers
- D׊:46000×93 mm
Specifications
About this item
The Tork conventional toilet roll provides the comfort of being at home, away from home. Tork Coreless Conventional toilet roll advanced balances cost and performance, and is suitable for low-traffic washrooms.
Reduce refills and cut maintenance time with this extra long toilet roll
- Eliminate waste, thanks to our unique coreless roll
- Impress guests with the attractive design of our 2-ply toilet paper
- Sheet length 115 mm
The Tork Easy Handling® plastic bag makes carrying, opening and disposal easier. This extra long roll lasts longer, reducing the frequency of refilling and providing superb value. Plus, its coreless design eliminates waste, leaving nothing to dispose of when the roll is finished.
Certifications:
EU Ecolabel | SE/004/001. FSC Recycled credit | SA-COC-008266.