Zadání objednávky
Czech Republic
ContactUsLinkComponent
Anti-VHH Rabbit Polyclonal Antibody
Anti-VHH Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-95273-100
Supplier:  Novus Biologicals
New Product
Anti-VHH Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-95273-100
Supplier:  Novus Biologicals
New Product

Specifications

  • Druh protilátky:
    Sekundární
  • Název antigenu:
    Anti-Llama
  • Symbol antigenu:
    VHH
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Hostitel:
    Rabbit
  • Izotyp:
    IgG
  • Western blot:
    Yes
  • Synonyma antigenu:
    anti-VhH Antibody|camelid VhH Antibody|single-domain antibody (sdAb)
  • Čistota:
    Multi-step
  • Skladovací pufr:
    0,02 M potassium phosphate, 0,15 M sodium chloride, pH 7,2
  • Skladovací teplota:
    Store at −20 °C short term. Aliquot and store at −80 °C long term. Avoid freeze-thaw cycles.
  • Imunogen:
    Rabbit anti-Llama VHH secondary antibody was prepared from whole rabbit serum produced by repeated immunizations with a VhH camelid domain protein
  • Rozměry protilátky:
    100 µl
  • Typ balení:
    Vial

Specifications

About this item

Recommended dilutions: ELISA 1:10000 to 1:50000; Western blot 1:1000 to 1:5000.

Specificity: Anti-VHH is monospecific antiserum processed by delipidation and defibrination followed by sterile filtration. Anti-Vhh antibody detects recombinant VHH proteins, native Llama IgG₂ and native Llama IgG₃.

Preservative: 0,01% sodium azide

Type: Secondary
Antigen: VHH 
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Llama