Tork Xpress® H2 Compressed Soft Multifold Hand Towels, White
Catalog # SCAA600454
Supplier: Essity
New Product
Specifications
- Určeno pro:Compatible with Tork H2 dispensers
- Balené:6 packs, 200 paper hand towels each; 2 ply
- Barva:Bílá
- Popis:Advanced multifold hand towels, refill
- D׊:80×212 mm (Folded size); 322×212 mm (Unfolded size)
Specifications
About this item
Tork Xpress® compressed soft multifold hand towels white H2, advanced, 2-ply, M-fold, large. Improve guest comfort with these strong, absorbent and large hand towels.
Reduce paper towel consumption in washrooms and improve hygiene with one-at-a-time dispensing
- Balance fantastic performance with great value, thanks to these economical hand paper towels
- Maximise your space - use these compressed paper hand towels with the compact Tork Xpress® dispenser in low-to-medium traffic washrooms
- Advanced quality, compatible with H2 dispensers
Increase capacity in smaller dispensers with 50% compressed paper hand towels. These large paper towels are ultra-absorbent and tear-resistant, so guests use fewer sheets. One-at-a-time dispensing helps reduce waste and improve hygiene, as guests only touch what they take. The EU Ecolabel and FSC certifications demonstrate your commitment to sustainability.
Certifications:
FSC Mix Virgin and Rec.fibre, SA-COC-008266; EU Ecolabel, SE/004/001.