NANOCOLOR® Photometric Tube Test Kits for Cyanide
Supplier: MACHEREY-NAGEL
Certificates
About this item
Photometric test kits for the determination of cyanide in water samples.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
- Precisely pre-dosed reagents with constant high quality from batch to batch
- Coloured pictogram instructions and easy handling of the tubes
- 16 mm (OD) tubes for easy pipetting
The test kits are evaluated in 16 mm (OD) tubes. They are preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Delivery: Rugged box with 20 test tubes.
Specifications
- Model:MANA985025
- Plošina:NANOCOLOR® tube tests
- Skladovací teplota:15…25 °C
- Životnost police (měsíce):12
- Hmotnost:624.1 g (including packaging)
Specifications
Related Information
Certificates
Search by Lot NumberCyanide 08 Tube Test .pdf NANOCOLOR® Cyanide Photometric Tube Tests.pdf
Additional Product Details
Frequently Bought Together