O tomto produktu
These tubes are low DNA binding, ensuring minimal sample loss, and free from PCR inhibitors, UV leachables, and human DNA, making them ideal for sensitive applications. Furthermore, the sustain series are sustainable - with biobased resin and ISCC certification, underscoring their eco-friendly design. The standard DNA low binding tubes ensure minimal sample loss, and free from PCR inhibitors, UV leachables, and human DNA, making them ideal for sensitive applications.
Low DNA binding
- PCR inhibitor free
- UV leachables free
- Human DNA free
-
Sustain series are sustainable with biobased resin and ISCC certification
Dimensions: 356×254×635 mm
Cap type: Snap cap
Ensuring that maximum DNA recovery is achieved and reducing the risk of sample loss and contamination. Particularly vital in applications such as PCR, sequencing, and cloning, where even small losses can significantly impact the results. Additionally, the assurance that these tubes are free from PCR inhibitors, UV leachables, and human DNA further enhances the reliability of experimental outcomes by preventing any potential interference with sensitive assays.
Specifikace
- Catalog No:
- 525-1625
- 525-1624
- 525-1626
- 525-1622
- 525-1623
- 525-1627
- Environmentally Preferable:
- Udržitelné materiály, Low Manufacturing Impact
- Udržitelné materiály, Low Manufacturing Impact
- Udržitelné materiály, Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certifikát ochrany životního prostředí:
- IRECS, ISCC PLUS - certified
- IRECS, ISCC PLUS - certified
- IRECS, ISCC PLUS - certified
- IRECS
- IRECS
- IRECS
Specifikace
Související informace
DNA Low Binding Snap Cap Microcentrifuge Tubes, Sustain Series.pdf
Další podrobnosti o produktu
TDS Low DNA Binding Snap Cap Microcentrifuge Tube.pdf TDS Snap Cap Microcentrifuge Tubes.pdf
Technický list
Thermo Scientific Pure Protection Microcentrifuge Tubes.pdf