Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
 

Human Recombinant p53 (from E. coli)

1286
  ABCAAB237007-100
 :   Abcam
Human Recombinant p53 (from E. coli)
  ABCAAB237007-100
 :  Abcam
 :  
Human Recombinant p53 (from E. coli)
Human Recombinant p53 (from E. coli)

1286

 :  
 
17 850,00 Kč
   
1 * 100 µG
 

Recombinant Human p53 protein is a Human Full Length protein in the 1 to 393 aa range with >90% purity and suitable for SDS-PAGE and mass spectrometry. The predicted molecular weight of ab237007 protein is 59,7 kDa. Save time and ensure accurate results. Use our p53 protein as a control .

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

 

 

  • Sekvence:
    1286
  • Zdroj:
    E. coli
  • Biologická aktivita:
    Standard
  • Čistota:
    >90%
  • Podmínky uskladnění:
    Store at –20 °C
  • Formulace:
    pH: 7,2 - 7,4, Constituents: Tris buffer, 50% Glycerol (glycerin, glycerine)
  • Druhy:
    Human
  • Cat. No.:
    ABCAAB237007-100
  • Transportní teplota:
    20
  • Testované aplikace:
    MS, SDS-PAGE
  • Formulář:
    Liquid
  • Bez obsahu živočichů:
    N
  • Název proteinu/peptidu:
    p53
  • Proteinová synonyma:
    P53|TP53|Cellular tumor antigen p53|Antigen NY-CO-13|Phosphoprotein p53|Tumor suppressor p53
  • Typ proteinu/peptidu:
    Recombinant
  • Spojení:
    Unconjugated

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.