Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Recombinant HPV16 E6 (from E. coli)

Recombinant HPV16 E6 (from E. coli)

1286
Katalogové číslo ABCAAB226447-100
Marketsource
Dodavatel:   Abcam
Recombinant HPV16 E6 (from E. coli)
Katalogové číslo ABCAAB226447-100
Dodavatel:  Abcam
Marketsource
Číslo CAS:  
Recombinant HPV16 E6 (from E. coli)
Recombinant HPV16 E6 (from E. coli)

1286

Přejít na:
Číslo CAS:  
Price
17 850,00 Kč
Přihlásit se pro dostupnost
1 * 100 µG
Product Details & Documents

Recombinant HPV16 E6 protein (His tag) is a Human papillomavirus type 16 E6 Full Length protein, in the 1 to 158 aa range with >90% purity and suitable for SDS-PAGE and mass spectrometry. The predicted molecular weight of ab226447 protein is 23,2 kDa. Save time and ensure accurate results - use HPV16 E6 protein as a control.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikace

Specifikace

  • Sekvence:
    1286
  • Zdroj:
    E. coli
  • Biologická aktivita:
    Standard
  • Čistota:
    >90%
  • Podmínky uskladnění:
    Store at −20 °C
  • Formulace:
    pH: 7,2 - 7,4; Constituents: Tris buffer, 50% Glycerol (glycerin, glycerine)
  • Cat. No.:
    ABCAAB226447-100
  • Transportní teplota:
    20
  • Testované aplikace:
    MS, SDS-PAGE
  • Formulář:
    Liquid
  • Bez obsahu živočichů:
    N
  • Název proteinu/peptidu:
    HPV16 E6
  • Proteinová synonyma:
    Protein E6|E6
  • Typ proteinu/peptidu:
    Recombinant
  • Spojení:
    Unconjugated

Specifikace