Mouse MCP1 Matched Antibody Pair Kit
ABCAAB210894-5X96
: Abcam
:
- Popis:Mouse MCP1 Matched Antibody Pair Kit
- Testů v kitu:5×96 tests
Mouse MCP1 Matched Antibody Pair Kit is a kit for ELISA development for the measurement of Mouse MCP1 by sandwich ELISA method.