Rat Lumican ELISA Kit
Catalog # ABCAAB213917-96
Supplier: Abcam
CAS Number:
Specifications
- Popis:Rat Lumican ELISA Kit
- Testů v kitu:96 tests
Specifications
About this item
Rat Lumican ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat Lumican in Rat in Cell culture supernatant, Heparin Plasma, Serum samples.