ContactUsLinkComponent
Rat Lumican ELISA Kit
Rat Lumican ELISA Kit
Catalog # ABCAAB213917-96
Supplier:  Abcam
CAS Number:  
undefined
Rat Lumican ELISA Kit
Catalog # ABCAAB213917-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Popis:
    Rat Lumican ELISA Kit
  • Testů v kitu:
    96 tests
Specifications
About this item
Rat Lumican ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat Lumican in Rat in Cell culture supernatant, Heparin Plasma, Serum samples.