Mouse ACE2 ELISA Kit
Katalogové číslo ABCAAB213843-96
Dodavatel: Abcam
Číslo CAS:
Specifikace
- Popis:Mouse ACE2 ELISA kit
Specifikace
O tomto produktu
Mouse ACE2 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse ACE2 in Mouse in cell culture supernatant, Heparin plasma, serum samples.