Human NOV ELISA Kit (Nephroblastoma Overexpressed)
Catalog # ABCAAB193710-96
Supplier: Abcam
CAS Number:
Specifications
- Popis:Human NOV ELISA Kit (Nephroblastoma Overexpressed)
- Testů v kitu:96 tests
Specifications
About this item
Human NOV ELISA Kit (Nephroblastoma Overexpressed) is a Sandwich (quantitative) ELISA kit for the measurement of Human NOV (Nephroblastoma Overexpressed) in Human in Plasma, Cell culture supernatant, Serum samples.