Mouse Apolipoprotein AI Matched Antibody Pair Kit
Katalogové číslo ABCAAB221440-5X96
Dodavatel: Abcam
Číslo CAS:
Specifikace
- Popis:Mouse Apolipoprotein AI Matched Antibody Pair kit
Specifikace
O tomto produktu
Mouse Apolipoprotein AI Matched Antibody Pair kit is a kit for ELISA development for the measurement of Mouse Apolipoprotein AI by sandwich ELISA method.