ContactUsLinkComponent
MitoBiogenesis™ In-Cell ELISA Kit (Fluorescent)
MitoBiogenesis™ In-Cell ELISA Kit (Fluorescent)
Katalogové číslo ABCAAB140359-96
Dodavatel:  Abcam
Číslo CAS:  
undefined
MitoBiogenesis™ In-Cell ELISA Kit (Fluorescent)
Katalogové číslo ABCAAB140359-96
Dodavatel:  Abcam
Číslo CAS:  
Specifikace
  • Popis:
    MitoBiogenesis™ In-Cell ELISA kit (Fluorescent)
  • Skladovací teplota:
    +4 ℃
  • Testů v kitu:
    96 tests
Specifikace
O tomto produktu
MitoBiogenesis™ In-cell ELISA kit (Fluorescent) is cell-based (quantitative) ELISA for the measurement of MitoBiogenesis™ (Fluorescent) production in Mouse, Rat, Cow, Human in cell culture extracts samples.