Canakinumab ELISA Kit test
Catalog # ABCAAB282896-1X96
Supplier: Abcam
CAS Number:
Specifications
- Popis:Canakinumab ELISA kit test
Specifications
About this item
Canakinumab ELISA kit test is a Sandwich (quantitative) ELISA kit for the measurement of Canakinumab test in Human in plasma, serum samples.