Mouse M-CSF ELISA Kit
ABCAAB155457-96
: Abcam
:
- Popis:Mouse M-CSF ELISA kit
- Testů v kitu:96 tests
Mouse M-CSF ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse M-CSF in Mouse in Plasma, Cell culture supernatant, Serum samples.