profi ergo ENB20A Safety Gloves, 60147 Series
Katalogové číslo UVEX6014705
Dodavatel: UVEX
Specifikace
- Velikost rukavic:11
- Obouruké/jednoruké:Jednoruké
- Jednorázové/znovupoužitelné:Jednorázové
- Pudrované/nepudrované:Nepudrované
- Barva:Bílá/oranžová
- Norma EN:EN 420:2003 + A1:2009/EN 388:2016 + A1:2018/EN 407:2020
Specifikace
O tomto produktu
The thick special NBR coating of the profi ergo glove is resistant to oil and grease.
- Very good grip in wet and oily areas
- Coating on palm and parts of the back of the hand
- Suitable for operating touch displays
- Food-safe
The high-quality cotton interlock lining provides high wearer comfort, feels pleasant against the skin, and provides optimum regulation of the glove climate. The glove is also suitable for operating touch displays.