ContactUsLinkComponent
profi ergo ENB20A Safety Gloves, 60147 Series
profi ergo ENB20A Safety Gloves, 60147 Series
Katalogové číslo UVEX6014705
Dodavatel:  UVEX
profi ergo ENB20A Safety Gloves, 60147 Series
Katalogové číslo UVEX6014705
Dodavatel:  UVEX
Specifikace
  • Velikost rukavic:
    11
  • Obouruké/jednoruké:
    Jednoruké
  • Jednorázové/znovupoužitelné:
    Jednorázové
  • Pudrované/nepudrované:
    Nepudrované
  • Barva:
    Bílá/oranžová
  • Norma EN:
    EN 420:2003 + A1:2009/EN 388:2016 + A1:2018/EN 407:2020
Specifikace
O tomto produktu
The thick special NBR coating of the profi ergo glove is resistant to oil and grease.
  • Very good grip in wet and oily areas
  • Coating on palm and parts of the back of the hand
  • Suitable for operating touch displays
  • Food-safe
The high-quality cotton interlock lining provides high wearer comfort, feels pleasant against the skin, and provides optimum regulation of the glove climate. The glove is also suitable for operating touch displays.