profi ergo XG20 Safety Gloves, 60208 Series
Catalog # UVEX6020811
Supplier: UVEX
Specifications
- Velikost rukavic:11
- Obouruké/jednoruké:Jednoruké
- Jednorázové/znovupoužitelné:Jednorázové
- Pudrované/nepudrované:Nepudrované
- Barva:Black/orange/white
- Norma EN:EN ISO 21420:2020/EN 388:2016 + A1:2018/EN 407:2020
Specifications
About this item
The profi ergo XG safety glove range features an innovative Xtra grip coating made from nitrile rubber. It combines protection and grip with outstanding wearer comfort and flexibility.
- Fully coated back of hand
- Xtra Grip Technology for outstanding grip
- Excellent wearer comfort
- Multilayer design provides an outstanding service life
The profi ergo series are characterised by very good grip in damp, wet, and oily areas, good tactile feel, ergonomic fit, high flexibility, and particularly good wearer comfort due to the high moisture absorption properties of the cotton lining. The palm and entire back of the hand are coated with special NBR and Xtra Grip coatings (nitrile rubber). Free of harmful substances in accordance with OEKO-TEX® Standard 100.