Anti-MEK1 + MEK2 Rabbit Monoclonal Antibody [clone: EPR16667] (HRP (Horseradish Peroxidase))
Catalog # ABCAAB200179-100
Supplier: Abcam
Specifications
- Sekvence:1286
- Rozměry protilátky:100 µl
- Typ balení:Vial
Specifications
About this item
HRP Rabbit monoclonal [EPR16667] to MEK1 + MEK2.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: MEK1 + MEK2
Clonality: Monoclonal
Clone: EPR16667
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat