Order Entry
Czech Republic
ContactUsLinkComponent
Anti-G3BP Rabbit Monoclonal Antibody [clone: EPR13985(B)] (APC)
Anti-G3BP Rabbit Monoclonal Antibody [clone: EPR13985(B)] (APC)
Catalog # ABCAAB303138-100
Supplier:  Abcam
Anti-G3BP Rabbit Monoclonal Antibody [clone: EPR13985(B)] (APC)
Catalog # ABCAAB303138-100
Supplier:  Abcam

Specifications

  • Sekvence:
    1286
  • Rozměry protilátky:
    100 µl
  • Typ balení:
    Vial

Specifications

About this item

APC Rabbit monoclonal [EPR13985(B)] to G3BP.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: G3BP
Clonality: Monoclonal
Clone: EPR13985(B)
Conjugation: APC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human