ContactUsLinkComponent
Anti-GCDFP 15 Rabbit Monoclonal Antibody [clone: EP1582Y] (PE (Phycoerythrin))
Anti-GCDFP 15 Rabbit Monoclonal Antibody [clone: EP1582Y] (PE (Phycoerythrin))
Katalogové číslo ABCAAB303193-100
Dodavatel:  Abcam
Anti-GCDFP 15 Rabbit Monoclonal Antibody [clone: EP1582Y] (PE (Phycoerythrin))
Katalogové číslo ABCAAB303193-100
Dodavatel:  Abcam
O tomto produktu
PE Rabbit monoclonal [EP1582Y] to GCDFP 15.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: GCDFP 15
Clonality: Monoclonal
Clone: EP1582Y
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: