Anti-eIF4EBP1 Rabbit Monoclonal Antibody [clone: YCA-R330-40-8-6Â H3L2]
Dodavatel: Abcam
Certifikáty
O tomto produktu
Frequently Bought Together
VWR®, Pipety Pasteurovy, Standard Line
Dodavatel: VWR Collection
VWR® Ultra-High Performance Centrifuge Tubes
Dodavatel: VWR Collection
VWR®, Sáčky do homogenizátoru
Dodavatel: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Dodavatel: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Dodavatel: EPPENDORF
VWR®, Centrifuge Tubes, Conical Bottom
Dodavatel: VWR Collection
VWR®, Sérologická pipeta, standardní
Dodavatel: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Dodavatel: VWR Collection
Round-bottom polystyrene tubes, disposable, Falcon®
Dodavatel: Corning
VWR®, Nádoby na vzorky se šroubovacím uzávěrem
Dodavatel: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
Dodavatel: VWR Collection
VWR® Water Sampling Bottles, with Tamper Evident Cap
Dodavatel: VWR Collection
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection
VWR®, Kryoboxy, lepenka, 133×133 mm
Dodavatel: VWR Collection