Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Wnt16 Rabbit Recombinant Multiclonal Antibody [clone: RP23040122]
  ABCAAB308129-100UG
 :  Abcam
undefined
Anti-Wnt16 Rabbit Recombinant Multiclonal Antibody [clone: RP23040122]
  ABCAAB308129-100UG
 :  Abcam

 

  • Sekvence:
    1286
  • Rozměry protilátky:
    100 µg
  • Typ balení:
    Vial

 

 

Rabbit recombinant multiclonal [RP23040122] to Wnt16.

 : All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Wnt16
Clonality: Recombinant Multiclonal
Clone: RP23040122
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse