Anti-Histone H3 (di methyl K4) Rabbit Monoclonal Antibody [clone: Y47] (Alexa Fluor® 405)
Katalogové číslo ABCAAB207865-100
Dodavatel: Abcam
Specifikace
- Rozměry protilátky:100 µl
- Typ balení:Vial
Specifikace
O tomto produktu
Alexa Fluor® 405 Rabbit monoclonal [Y47] to Histone H3 (di methyl K4).
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: Y47
Conjugation: Alexa Fluor® 405
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human