Micro burner with needle valve, cartridge type
Catalog # USBE1423
Supplier: USBECK Laborgeräte
Specifications
- Max. tepl.:1640 °C
- Popis:Micro burner, without cartridge
- Bal.:1
Specifications
About this item
Micro burner with piezo electric ignition and air regulation. Maximum temperature of 1640 °C.