Cartridge burner, Teclu
Catalog # 451-0177
Supplier: USBECK Laborgeräte
Specifications
- Max. tepl.:1640 °C
- Popis:Bunsen burner with needle valve, without cartridge
- Bal.:1
Specifications
About this item
Cartridge burner with air regulation. Maximum temperature of 1640 °C.