Reagent/centrifuge tubes, round bottom, with screw cap
Catalog # SARS62.610.201
Supplier: SARSTEDT
Specifications
- Typ uzávěru:Šroubovací víčko
- Ø×D:15,3×92 mm
- Balené:500/case
- Verze:assembled cap, sterile, writing area
- Kapacita:10 ml
- Sekvence:10
- Barva víčka:Přírodní
- Bal.:500
- Max. RCF:3000×g
Specifications
About this item
PP, with HDPE screw cap or PP/PS with PP screw cap.
Universal tubes for a wide range of applications.