Síran vápenatý, Drierite™ 10-20 mesh for drying
Catalog # 43058.TD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:454 g
- Sekvence:3635
Specifications
About this item
:
Merck Index:14,1706
Melting Pt:1450 °C
Density:2,96
EINECS:231-900-3
:GHS07
Storage Temperature:Ambient
CAS Number:7778-18-9
Formula:CaO₄S
MDL Number:MFCD00010912