Orders LinkContactUsLinkComponent
Automatic burettes, Pellet pattern, class AS
Automatic burettes, Pellet pattern, class AS
Dodavatel:   SciLabware
Automatic burettes, Pellet pattern, class AS
Dodavatel:  SciLabware
Certifikáty
Automatic burettes, Pellet pattern, class AS
Automatic burettes, Pellet pattern, class AS
3 Dostupných variant níže
Product Details & Documents
Burettes made of chemically resistant PYREX® borosilicate glass.
  • Automatic zero facility
  • Simple operation
  • White enamel graduations
  • 1,5 mm bore PTFE key stopcock
Certifications: Complies with ISO 385 and DIN 12700.
Product Family Options
Informace o produktu
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Blowing ball, rubber, for automatic burettes, Pellet pattern
    Blowing ball, rubber, for automatic burettes, Pellet pattern
    BIBB3297/14AS
    02
    Přihlásit se pro dostupnost
    1 * 1 KS - 841,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Reservoir bottle, glass, clear, 2000 ml, with 29/32 joint
    Reservoir bottle, glass, clear, 2000 ml, with 29/32 joint
    BIBB3297/12
    01
    Přihlásit se pro dostupnost
    1 * 1 KS - 1 740,00 Kč