General-purpose thermometer
01, −10…+250 °C, NS 14/23, Blue spirit
Katalogové číslo 620-2290
Dodavatel: SciLabware
Přejít na:
Product Details & Documents
Spirit thermometers.
An integral ground glass cone on the stem allows direct fitting into Quickfit® jointed apparatus. Spirit thermometers have blue or green spirit for improved visibility.
Specifikace
- Sekvence:01
- Dělení:NS 14/23
- Náplň:Blue spirit
- Rozsah:−10…+250 °C