Analogue stopwatch
Stopwatch, 0 - 30 min
Katalogové číslo HECH41978010
Dodavatel: GLASWARENFABRIK KARL HECHT
Přejít na:
Product Details & Documents
Solid metal stopwatch with 7 jewels.
- Graduation 0 to 60 in 1/5 seconds and 1/100 minutes
- Timing capacity: Up to 30 minutes
- Impact, dust and water resistant
Specifikace
- Popis:Stopwatch, 0 - 30 min