Conductive pipette tips
GREI686288
- Balené:60/F-rack
- Sterilní:+
- Objem:200 - 1000 µl
- Environmentally Preferable:
- Druh špičky:Conductive, with ultra micro filter
- Barva špičky:Black
- Filtr:+
- Bal.:600
Conductive tips are used to prevent carryover in automated pipetting robots. The addition of graphite to the raw material PP makes the pipette tips electrically conductive. By measuring electric currents, the filling level can be determined in sample and reagent containers, so that the depth of immersion of the tip can be adjusted to the filling level.
- Filter tip: Free of detectable DNase, RNase, human DNA; non pyrogenic, non cytotoxic