Kieselguhr filter papers
Dodavatel: MACHEREY-NAGEL
Certifikáty
O tomto produktu
This filter paper retains very fine turbidities and is e.g. recommended for the clarification of urines or sugar solutions.
Specifikace
- Catalog No:
- MANA447012
- MANA547015
- MANA447015
- MANA447009
- MANA447011
- MANA447018
- MANA447032
- MANA547032
- MANA447024
- MANA547018
- MANA547024
- MANA547011
- MANA547012
- Hmotnost:
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- Tloušťka:
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- Rychlost filtrace:
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)