Solid phase extraction cartridges, Bond Elut™ PPL
O tomto produktu
Bond Elut™ PPL is a styrene-divinylvenzene (SDVB) polymer that has been modified with a proprietary non-polar surface. PPL will retain even the most polar classes of analytes, including phenols. The large particle size allows ease of flow for viscous or particulate rich water samples, while the high surface area and strong hydrophobicity ensure reproducible extractions with high recoveries upon elution. Bond Elut™ PPL is suitable for EPA Method 528 ‘Determination of Phenols in Drinking Water by SPE and Capillary GC/MS’.
- Modified styrene-divinylbenzene polymer
- Large particle size allows fast extraction speeds
- High surface area and capacity for polar analytes
9 Dostupných variant níže
Product Details & Documents
Bond Elut™ PPL is a styrene-divinylvenzene (SDVB) polymer that has been modified with a proprietary non-polar surface. PPL will retain even the most polar classes of analytes, including phenols. The large particle size allows ease of flow for viscous or particulate rich water samples, while the high surface area and strong hydrophobicity ensure reproducible extractions with high recoveries upon elution. Bond Elut™ PPL is suitable for EPA Method 528 ‘Determination of Phenols in Drinking Water by SPE and Capillary GC/MS’.
- Modified styrene-divinylbenzene polymer
- Large particle size allows fast extraction speeds
- High surface area and capacity for polar analytes
Specifications for Product Family
Specifikace
- Catalog No:
- 551-0331
- VARI12105003
- 551-0332
- 551-0333
- VARI12105006
- 551-0400
- VARI12102148
- 551-0401
- VARI12256087
- Povrchová oblast:
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
Specifikace
Product Family Options
Informace o produktu
- SekvenceFázeVelikost pórůVelikost částicKapacitaVýška vrstvyDostupnostPrice
- 551-0331Straight barrel cartridgeStraight barrel cartridge01PPL150 Å125 µm1 ml50 mgPřihlásit se pro dostupnost1 * 100 KS9 207,00 KčSpecifikace:
More
Specifications
Cat. No.551-0331Povrchová oblast600 m²/g - VARI12105003Straight barrel cartridgeStraight barrel cartridge02PPL150 Å125 µm1 ml100 mgPřihlásit se pro dostupnost1 * 100 KS11 815,65 KčSpecifikace:
More
Specifications
Cat. No.VARI12105003Povrchová oblast600 m²/g - 551-0332Straight barrel cartridgeStraight barrel cartridge03PPL150 Å125 µm3 ml100 mgPřihlásit se pro dostupnost1 * 50 KS7 877,10 KčSpecifikace:
More
Specifications
Cat. No.551-0332Povrchová oblast600 m²/g - 551-0333Straight barrel cartridgeStraight barrel cartridge04PPL150 Å125 µm3 ml200 mgPřihlásit se pro dostupnost1 * 50 KS10 884,72 KčSpecifikace:
More
Specifications
Cat. No.551-0333Povrchová oblast600 m²/g - VARI12105006Straight barrel cartridgeStraight barrel cartridge05PPL150 Å125 µm3 ml500 mgPřihlásit se pro dostupnost1 * 50 KS16 009,95 KčSpecifikace:
More
Specifications
Cat. No.VARI12105006Povrchová oblast600 m²/g - 551-0400Straight barrel cartridgeStraight barrel cartridge06PPL150 Å125 µm6 ml500 mgPřihlásit se pro dostupnost1 * 30 KS11 232,54 KčSpecifikace:
More
Specifications
Cat. No.551-0400Povrchová oblast600 m²/g - VARI12102148Straight barrel cartridgeSpecifikace:
More
Specifications
Cat. No.VARI12102148Povrchová oblast600 m²/g - 551-0401Straight barrel cartridgeSpecifikace:
More
Specifications
Cat. No.551-0401Povrchová oblast600 m²/g - VARI12256087Straight barrel cartridgeSpecifikace:
More
Specifications
Cat. No.VARI12256087Povrchová oblast600 m²/g