Filter forceps
Straight, blunt
Katalogové číslo MILYXX6200006P
Dodavatel: MILLIPORE
Přejít na:
Product Details & Documents
Sterilisable filter forceps made from stainless steel, with PP covered handles (in three different colours).
Specifikace
- Type:Straight, blunt