ContactUsLinkComponent
UV transilluminators, slimline, single wavelength, for use with Syngene InGenius and U:Genius systems
UV transilluminators, slimline, single wavelength, for use with Syngene InGenius and U:Genius systems
Katalogové číslo 733-2310
undefined
UV transilluminators, slimline, single wavelength, for use with Syngene InGenius and U:Genius systems
Katalogové číslo 733-2310
Specifikace
  • Model:
    GE-2020M-E
  • Vlnová délka:
    302 nm
  • Popis:
    UV transilluminator, with runners
  • Sekvence:
    01
  • Velikost filtru:
    200×200 mm
  • Bal.:
    1