Solid phase extraction cartridges, Bond Elut™ CH
Dodavatel: VARIAN
O tomto produktu
Bond Elut™ CH is a mid-polarity sorbent that exhibits unique selectivities for certain analytes. When employed as a non-polar sorbent, CH has the approximate polarity of a C₂ sorbent. Bond Elut™ CH is often a good choice when non-polar sorbents such as C₁₈, C₈ or C₂ do not provide the desired selectivity.
- Non-polar CH with polarity similar to C₂
- Retains polar analytes from aqueous matrices
- Good choice when common non-polar sorbents do not provide the required selectivity
5 Dostupných variant níže
Product Details & Documents
Bond Elut™ CH is a mid-polarity sorbent that exhibits unique selectivities for certain analytes. When employed as a non-polar sorbent, CH has the approximate polarity of a C₂ sorbent. Bond Elut™ CH is often a good choice when non-polar sorbents such as C₁₈, C₈ or C₂ do not provide the desired selectivity.
- Non-polar CH with polarity similar to C₂
- Retains polar analytes from aqueous matrices
- Good choice when common non-polar sorbents do not provide the required selectivity
Specifications for Product Family
Specifikace
- Catalog No:
- VARI12113032
- VARI12102006
- VARI12102033
- VARI12256005
- HEWL12256039
- Naplnění uhlíkem:
- 9,6%
- 9,6%
- 9,6%
- 9,6%
- 9,6%
- S navazanou skupinou:
- Yes
- Yes
- Yes
- Yes
- Yes
- Povrchová oblast:
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
Specifikace
Product Family Options
LRC cartridges
Informace o produktu
- SekvenceFázeVelikost pórůKapacitaVýška vrstvyDostupnostPrice
- VARI12113032LRC cartridgeSpecifikace:
More
Specifications
Cat. No.VARI12113032Naplnění uhlíkem9,6%S navazanou skupinouYesPovrchová oblast500 m²/g
Straight barrel cartridges
Informace o produktu
- SekvenceFázeVelikost pórůKapacitaVýška vrstvyDostupnostPrice
- VARI12102006Straight barrel cartridgeSpecifikace:
More
Specifications
Cat. No.VARI12102006Naplnění uhlíkem9,6%S navazanou skupinouYesPovrchová oblast500 m²/g - VARI12102033Straight barrel cartridgeSpecifikace:
More
Specifications
Cat. No.VARI12102033Naplnění uhlíkem9,6%S navazanou skupinouYesPovrchová oblast500 m²/g - VARI12256005Straight barrel cartridgeSpecifikace:
More
Specifications
Cat. No.VARI12256005Naplnění uhlíkem9,6%S navazanou skupinouYesPovrchová oblast500 m²/g - HEWL12256039Straight barrel cartridgeSpecifikace:
More
Specifications
Cat. No.HEWL12256039Naplnění uhlíkem9,6%S navazanou skupinouYesPovrchová oblast500 m²/g