Orders LinkContactUsLinkComponent
General purpose sample storage vials and caps
General purpose sample storage vials and caps
Supplier:   Agilent
General purpose sample storage vials and caps
Supplier:  Agilent
Certificates
General purpose sample storage vials and caps
General purpose sample storage vials and caps
5 Options Available Below
Product Details & Documents
Product Family Options
General purpose sample storage vial kits
Product Information
  • Sekvence
    Kapacita
    Ø×H
    Barva
    Čepice součástí
    Typ víčka
    Velikost víčka
    Materiál uzávěru
    Barva víčka
    Septum
    Availability
    Price
  • 01
    4,00 ml
    15×45 mm
    Průhledná
    Ano
    13-425
    PP
    PTFE/silicone
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS
  • 07
    22,00 ml
    23×85 mm
    Jantarová
    Ano
    20-400
    PP
    PTFE/silicone
    Log in for availability
    Log in for availability
    1 * 100 KS
    1 * 100 KS
  • 08
    40,00 ml
    28×95 mm
    Průhledná
    Ano
    24-414
    PP
    PTFE/silicone
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS
  • 10
    40,00 ml
    28×95 mm
    Jantarová
    Ano
    24-414
    PP
    PTFE/silicone
    Log in for availability
    Log in for availability
    1 * 100 KS
    1 * 100 KS
  • 11
    40,00 ml
    28×95 mm
    Jantarová
    Ano
    Open top
    24-414
    PP
    Bílá
    Log in for availability
    Log in for availability
    1 * 100 KS
    1 * 100 KS
Bonded caps
Product Information
  • Catalog #
    Materiál
    Sekvence
    Popis
    Velikost víčka
    Availability
    Price
  • PP
    04
    Bonded white caps with PTFE/silicone liner
    24-414
    Log in for availability
    Log in for availability
    1 * 100 KS
    1 * 100 KS - 0,00 Kč
    0,00 Kč
    1 * 100 KS
       0,00 Kč