Sharps disposal container, CLINIPAK
Catalog # LPIT138013
Supplier: LP ITALIANA
Specifications
- Ø×H:200×170 mm
- Kapacita:4 l
- Bal.:40
Specifications
About this item
Sharp disposal container, plastic, with special lid for the extraction of needles.