Orders LinkContactUsLinkComponent
Cell scrapers
Cell scrapers
Cell scraper, 50 mm
Catalog # WITG8.486.200
Supplier:   witeg Labortechnik
Cell scrapers
Catalog # WITG8.486.200
Supplier:  witeg Labortechnik
Cell scrapers
Cell scrapers
Cell scraper, 50 mm
Price
2 540,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Bent, plastic coated glass handle, mobile PTFE blade. Scraper for removal of cells from flasks and plates. Length 50 mm.
Specifications
  • Popis:
    Cell scraper
  • Celková délka:
    50 mm