Dropping bottles with stoppers
100 ml
Katalogové číslo WITG5.883.100
Dodavatel: witeg Labortechnik
Přejít na:
Product Details & Documents
Glass, amber.
- Good chemical resistance
- Amber coloured to protect light sensitive substances
Dropping bottle with grooved flat stopper.
Specifikace
- Kapacita:100 ml