Stopcocks for desiccators
Katalogové číslo WITG5.662.004
Dodavatel: witeg Labortechnik
Specifikace
- Popis:Stopcock with PTFE spindle, for desiccator lid outlet
- Bal.:1
Specifikace
O tomto produktu
Stopcock, with FCH-V 24/29, flat-free high vacuum joint, made of borosilicate glass.