Strips of 8 PCR tubes with attached domed individual caps
Catalog # BITX3728.8S.X
Supplier: BIOTIX
Specifications
- Balené:10 bags of 12/pack
- Kapacita:0,2 ml
- Barva:Průhledná
- Popis:strips of 8 PCR tubes, with attached domed individual caps
- Sekvence:05
- Bal.:120
Specifications
About this item
These PCR strip tubes are made of high quality virgin polypropylene and feature thin walls for efficient heat transfer.
- Translucent strip tubes with domed caps
- Thin walled tube for best heat transfer
- DNase, RNase and endotoxin free
Certifications: CE certified, IVD labeled and manufactured under stringent controls in an ISO 9001 certified facility. Every lot is tested for precision ensuring maximum quality and compatibility. All raw materials are traceable.