Verification standard for Spectroquant® Multy and Spectroquant® Move 100 colorimeters, Supelco®
Catalog # 1.19302.0001
Supplier: Merck
CAS Number:
Specifications
- Bal.:1
- Popis:Standardy Spectroquant® pro kolorimetry Spectroquant® Multy a Spectroquant® Move 100
Specifications
About this item
Designed to reassure the user about the accuracy and the reliability of Spectroquant® Multy and Spectroquant® Move 100 colorimeters.
Delivery: Set includes seven sealed cells including a zero cell and stable coloured solutions for measuring absorption at 430, 530, 560, 580, 610 and 660 nm, certificate and instructions.